OriGene
Manufacturer Code:TA503259
Catalog # 50-167-4199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 4-185 of human DHFRL1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DHFRL1 DHFRP4; Dihydrofolate reductase 2, mitochondrial; dihydrofolate reductase like 1; dihydrofolate reductase pseudogene 4; Dihydrofolate reductase, mitochondrial; dihydrofolate reductase-like 1; Dihydrofolate reductase-like protein 1
Gene Aliases: DHFR2; DHFRL1; DHFRP4
UniProt ID: (Human) Q86XF0
Entrez Gene ID: (Human) 200895
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.