Invitrogen
Manufacturer Code:MA112599
Catalog # 01-674-367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | T101 |
Immunogen | Recombit type 2 dihydrodiol dehydrogenase from human lung cancer |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-deoxyglucosone reductase; Can2DD; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; DDH; dihydrodiol dehydrogenase (dimeric); Dimeric dihydrodiol dehydrogenase; Hum2DD; putative LOC617564 protein; Sus2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; xylose dehydrogenase 1
Gene Aliases: 1300018L09Rik; 2DD; AA591799; CAN2DD; ch211-203b17.3; DHDH; HUM2DD; SUS2DD; XELAEV_18036112mg; Xld-1; Xld1; zgc:101723
UniProt ID: (Dog) Q9TV68, (Human) Q9UQ10, (Bovine) Q148L6, (Pig) Q9TV69, (Mouse) Q9DBB8, (Xenopus) Q6DKE0, (Zebrafish) Q642M9
Entrez Gene ID: (Rabbit) 100008610, (Dog) 403786, (Human) 27294, (Bovine) 617564, (Pig) 397337, (Rat) 691002, (Mouse) 71755, (Xenopus) 444539, (Zebrafish) 449558
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.