Invitrogen
Manufacturer Code:PA5113505
Catalog # PIPA5113505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Trans-12-dihydrobenzene-12-diol dehydrogenase protein (1-334AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300018L09Rik 2DD 3-deoxyglucosone reductase AA591799 DDH Dhdh dihydrodiol dehydrogenase dihydrodiol dehydrogenase (dimeric) Dimeric dihydrodiol dehydrogenase D-xylose 1-dehydrogenase D-xylose-NADP dehydrogenase Hum2DD Sus2DD Trans-12-dihydrobenzene-12-diol dehydrogenase Xld1 Xld-1 xylose dehydrogenase 1; 3-deoxyglucosone reductase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; dihydrodiol dehydrogenase (dimeric); Dimeric dihydrodiol dehydrogenase; Hum2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; xylose dehydrogenase 1
Gene Aliases: 1300018L09Rik; 2DD; AA591799; DHDH; HUM2DD; Xld-1; Xld1
UniProt ID: (Human) Q9UQ10, (Mouse) Q9DBB8
Entrez Gene ID: (Human) 27294, (Mouse) 71755
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.