Novus Biologicals
Manufacturer Code:NBP156950
Catalog # NBP156950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DHDH(dihydrodiol dehydrogenase (dimeric)) The peptide sequence was selected from the middle region of DHDH. Peptide sequence PCWCPTELVVKGEHKEFPLPPVPKDCNFDNGAGMSYEAKHVWECLRKGMK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-deoxyglucosone reductase; 3-deoxyglucosone reductase dihydrodiol dehydrogenase (dimeric) Dimeric dihydrodiol dehydrogenase D-xylose 1-dehydrogenase D-xylose-NADP dehydrogenase EC 1.1.1.1792DD EC 1.3.1.20 Hum2DD trans-12-dihydrobenzene-12-diol dehydrogenase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; dihydrodiol dehydrogenase (dimeric); Dimeric dihydrodiol dehydrogenase; Hum2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase
Gene Aliases: 2DD; DHDH; HUM2DD
UniProt ID: (Human) Q9UQ10
Entrez Gene ID: (Human) 27294
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.