Proteintech
Manufacturer Code:139001AP150UL
Catalog # 50-172-9077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DHDH Fusion Protein Ag4866 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2DD D xylose 1 dehydrogenase D xylose NADP dehydrogenase DHDH HUM2DD; 3-deoxyglucosone reductase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; dihydrodiol dehydrogenase (dimeric); Dimeric dihydrodiol dehydrogenase; Hum2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; xylose dehydrogenase 1
Gene Aliases: 1300018L09Rik; 2DD; AA591799; DHDH; HUM2DD; Xld-1; Xld1
UniProt ID: (Human) Q9UQ10, (Mouse) Q9DBB8
Entrez Gene ID: (Human) 27294, (Rat) 691002, (Mouse) 71755
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.