Invitrogen
Manufacturer Code:PA572414
Catalog # PIPA572414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7-dehydrocholesterol reductase; 7-dehydrocholesterol reductase 7-DHC reductase Putative sterol reductase SR-2 Sterol Delta(7)-reductase DHCR7 D7SR; 7-DHC reductase; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; putative sterol reductase SR-2; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2
Gene Aliases: AA409147; D7SR; DHCR7; SLOS
UniProt ID: (Human) Q9UBM7, (Bovine) Q5E9J5, (Mouse) O88455, (Rat) Q9Z2Z8
Entrez Gene ID: (Human) 1717, (Chicken) 422982, (Horse) 100061433, (Bovine) 514745, (Dog) 483675, (Mouse) 13360, (Rat) 64191
Molecular Function:
oxidoreductase
receptor
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.