Invitrogen
Manufacturer Code:PA5115116
Catalog # PIPA5115116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DHCR7 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7-dehydrocholesterol reductase; 7-dehydrocholesterol reductase 7-DHC reductase AA409147 D7SR delta-7-dehydrocholesterol reductase Delta7-sterol reductase Dhcr7 putative sterol reductase SR-2 SLOS sterol Delta(7)-reductase sterol delta-7-reductase Sterol reductase SR-2; 7-DHC reductase; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; putative sterol reductase SR-2; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2
Gene Aliases: AA409147; D7SR; DHCR7; SLOS
UniProt ID: (Human) Q9UBM7, (Mouse) O88455, (Rat) Q9Z2Z8
Entrez Gene ID: (Human) 1717, (Mouse) 13360, (Rat) 64191
Molecular Function: oxidoreductase receptor reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.