Invitrogen
Manufacturer Code:PA5113957
Catalog # PIPA5113957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DGKQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 110kDa DAG kinase theta DAGK DAGK4 DAGK7 Dgk theta Dgkd Dgkq DGK-theta Diacylglycerol kinase theta diacylglycerol kinase delta (110 kDa) diacylglycerol kinase theta diacylglycerol kinase theta 110kDa Diglyceride kinase theta testis tissue sperm-binding protein Li 38a; DAG kinase theta; DGK-theta; Diacylglycerol kinase theta; diacylglycerol kinase, theta 110kDa; Diglyceride kinase theta; testis tissue sperm-binding protein Li 38a
Gene Aliases: DAGK; DAGK4; DAGK7; DGKQ
UniProt ID: (Human) P52824
Entrez Gene ID: (Human) 1609
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.