Proteintech
Manufacturer Code:178851AP150UL
Catalog # 50-173-1854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DGKQ Fusion Protein Ag11938 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase theta; DAG kinase theta DAGK DAGK4 DAGK7 DGK theta DGKQ Diacylglycerol kinase theta Diglyceride kinase theta; Dgk theta; DGK-theta; Diacylglycerol kinase theta; diacylglycerol kinase, delta (110 kDa); diacylglycerol kinase, theta 110kDa; Diglyceride kinase theta; testis tissue sperm-binding protein Li 38a
Gene Aliases: 110kDa; DAGK; DAGK4; DAGK7; Dgkd; DGKQ
UniProt ID: (Human) P52824, (Rat) D3ZEY4, (Mouse) Q6P5E8
Entrez Gene ID: (Human) 1609, (Rat) 100361138, (Mouse) 110524
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.