Invitrogen
Manufacturer Code:PA542769
Catalog # PIPA542769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DGKH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase eta; DGK-eta; Diacylglycerol kinase eta; Diacylglycerol kinase eta DGKeta DKFZp761I1510; diacylglycerol kinase, eta; Diglyceride kinase eta
Gene Aliases: 5930402B05Rik; D130015C16; DGK; DGKeta; DGKH; RGD1561955; si:ch211-93a19.1
UniProt ID: (Human) Q86XP1, (Mouse) D3YXJ0
Entrez Gene ID: (Rabbit) 100341525, (Horse) 100060528, (Human) 160851, (Dog) 476932, (Mouse) 380921, (Guinea pig) 100736151, (Rat) 361076, (Zebrafish) 100137124
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.