Proteintech
Manufacturer Code:138731AP150UL
Catalog # 50-172-9057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DGKH Fusion Protein Ag4926 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase eta; DAG kinase eta DGK eta DGKeta DGKH Diacylglycerol kinase eta diacylglycerol kinase eta Diglyceride kinase eta; DGK-eta; Diacylglycerol kinase eta; diacylglycerol kinase, eta; Diglyceride kinase eta
Gene Aliases: 5930402B05Rik; D130015C16; DGK; DGKeta; DGKH; RGD1561955
UniProt ID: (Human) Q86XP1, (Mouse) D3YXJ0
Entrez Gene ID: (Human) 160851, (Mouse) 380921, (Rat) 361076
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.