Invitrogen
Manufacturer Code:PA553558
Catalog # PIPA553558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DGKE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase epsilon; DAGK5 DAGK6 DGK DAG kinase epsilon DGK-epsilon diacylglycerol kinase epsilon diglyceride kinase epsilon; DGK-epsilon; Diacylglycerol kinase epsilon; diacylglycerol kinase, epsilon 64kDa; Diglyceride kinase epsilon
Gene Aliases: AHUS7; DAGK5; DAGK6; DGK; DGKE; NPHS7
UniProt ID: (Human) P52429
Entrez Gene ID: (Human) 8526
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.