Invitrogen
Manufacturer Code:PA536250
Catalog # PIPA536250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 180-230 of human DGK- epsilon |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase epsilon; DGK-epsilon; Diacylglycerol kinase epsilon; Diacylglycerol kinase epsilon DAG kinase epsilon Diglyceride kinase epsilon DGK-epsilon DGKE DAGK5; diacylglycerol kinase, epsilon 64kDa; Diglyceride kinase epsilon
Gene Aliases: AHUS7; C87606; DAGK5; DAGK6; DGK; DGKE; NPHS7
UniProt ID: (Human) P52429, (Mouse) Q9R1C6
Entrez Gene ID: (Human) 8526, (Mouse) 56077, (Rat) 497978
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.