Invitrogen
Manufacturer Code:PA5101123
Catalog # PIPA5101123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DGKE corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AHUS7 C87606 DAG kinase epsilon DAGK5 DAGK6 DGK DGKE DGK-epsilon Diacylglycerol kinase epsilon diacylglycerol kinase epsilon diacylglycerol kinase epsilon 64kDa diglyceride kinase epsilon EC 2.7.1.107 NPHS7; DAG kinase epsilon; DGK-epsilon; Diacylglycerol kinase epsilon; diacylglycerol kinase, epsilon 64kDa; Diglyceride kinase epsilon
Gene Aliases: AHUS7; C87606; DAGK5; DAGK6; DGK; DGKE; NPHS7
UniProt ID: (Human) P52429, (Mouse) Q9R1C6
Entrez Gene ID: (Human) 8526, (Mouse) 56077, (Rat) 497978
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.