Abnova
Manufacturer Code:PAB30518
Catalog # 89-032-861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 63 - 192 of human DGKE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase epsilon; DAGK6; DGK; DGK-epsilon; Diacylglycerol kinase epsilon; diacylglycerol kinase, epsilon 64kDa; Diglyceride kinase epsilon
Gene Aliases: AHUS7; DAGK5; DAGK6; DGK; DGKE; NPHS7
UniProt ID: (Human) P52429
Entrez Gene ID: (Human) 8526
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.