Invitrogen
Manufacturer Code:PA575256
Catalog # PIPA575256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 34-88 of Human DGKD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa diacylglycerol kinase; DAG kinase delta; DGK-delta; Diacylglycerol kinase delta; Diacylglycerol kinase delta DAG kinase delta 130 kDa diacylglycerol kinase Diglyceride kinase delta DGK-delta DGKD KIAA0145 DGKdelta DGK delta; diacylglycerol kinase, delta; diacylglycerol kinase, delta 130kDa; Diglyceride kinase delta
Gene Aliases: AI841987; D330025K09; DGKD; dgkd-2; DGKdelta; KIAA0145; RGD1563309
UniProt ID: (Human) Q16760, (Mouse) E9PUQ8
Entrez Gene ID: (Human) 8527, (Mouse) 227333, (Rat) 368088
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.