Invitrogen
Manufacturer Code:PA545784
Catalog # PIPA545784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human DGKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa diacylglycerol kinase; DAG kinase delta; DGK-delta; Diacylglycerol kinase delta; diacylglycerol kinase DGKdelta FLJ26930 KIAA0145 dgkd-2; diacylglycerol kinase, delta; diacylglycerol kinase, delta 130kDa; Diglyceride kinase delta
Gene Aliases: AI841987; D330025K09; DGKD; dgkd-2; DGKdelta; KIAA0145; RGD1563309
UniProt ID: (Human) Q16760, (Mouse) E9PUQ8
Entrez Gene ID: (Human) 8527, (Rabbit) 100345378, (Dog) 486167, (Guinea pig) 100731863, (Mouse) 227333, (Rat) 368088
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.