Invitrogen
Manufacturer Code:PA5110941
Catalog # PIPA5110941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human DGKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa diacylglycerol kinase; 130 kDa diacylglycerol kinase AI841987 BOS_3869 D330025K09 DAG kinase delta DGK delta DGKD dgkd-2 DGKdelta DGK-delta Diacylglycerol kinase delta Diglyceride kinase Diglyceride kinase delta EC 2.7.1.107 FLJ26930 KIAA0145 RGD1563309; DAG kinase delta; DGK-delta; Diacylglycerol kinase delta; diacylglycerol kinase, delta 130kDa; Diglyceride kinase delta
Gene Aliases: DGKD; dgkd-2; DGKdelta; KIAA0145
UniProt ID: (Human) Q16760
Entrez Gene ID: (Human) 8527
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.