Abnova
Manufacturer Code:PAB3346
Catalog # 89-130-962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DGKD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DGKD.
For Research Use Only
Protein Aliases: 130 kDa diacylglycerol kinase; DAG kinase delta; DGK-delta; dgkd-2; DGKdelta; Diacylglycerol kinase delta; diacylglycerol kinase, delta 130kDa; Diglyceride kinase delta; FLJ26930; KIAA0145
Gene Aliases: DGKD; dgkd-2; DGKdelta; KIAA0145
UniProt ID: (Human) Q16760
Entrez Gene ID: (Human) 8527
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.