Invitrogen
Manufacturer Code:PA544058
Catalog # PIPA544058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DGKA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80 kDa diacylglycerol kinase; DAG kinase alpha; DGK-alpha; Diacylglycerol kinase alpha; diacylglycerol kinase alpha Flt2-Pr2; Diacylglycerol kinase DAGK DAGK1 DGK-alpha MGC12821 MGC42356; diacylglycerol kinase, alpha 80kDa; Diglyceride kinase alpha; truncated diacylglycerol kinase alpha Flt1-Pr1; truncated diacylglycerol kinase alpha Flt1-Pr2
Gene Aliases: 80kDa; AW146112; DAGK; DAGK1; DGK-alpha; DGKA
UniProt ID: (Human) P23743, (Pig) P20192, (Bovine) A0JN54, (Mouse) O88673, (Rat) P51556
Entrez Gene ID: (Rabbit) 100347223, (Horse) 100051662, (Human) 1606, (Pig) 397097, (Bovine) 506348, (Dog) 100855691, (Mouse) 13139, (Rat) 140866, (Guinea pig) 100721358
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.