Invitrogen
Manufacturer Code:PA5106475
Catalog # PIPA5106475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DGKA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80 kDa diacylglycerol kinase; 80 kDa diacylglycerol kinase 80kDa AW146112 DAG kinase alpha Dagk Dagk1 Dgka DGK-alpha Diacylglycerol kinase alpha diacylglycerol kinase alpha Flt2-Pr2 diacylglycerol kinase alpha diacylglycerol kinase alpha 80kDa Diglyceride kinase diglyceride kinase alpha MGC12821 MGC42356 truncated diacylglycerol kinase alpha Flt1-Pr1 truncated diacylglycerol kinase alpha Flt1-Pr2; DAG kinase alpha; DGK-alpha; Diacylglycerol kinase alpha; diacylglycerol kinase, alpha 80kDa; Diglyceride kinase alpha
Gene Aliases: DAGK; DAGK1; DGK-alpha; DGKA
UniProt ID: (Human) P23743
Entrez Gene ID: (Human) 1606
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.