Novus Biologicals
Manufacturer Code:NBP229920
Catalog # NBP229920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Clone | RB3716 |
Immunogen | This DGKZ antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 989-1019 amino acids from the C-terminal region of human DGKZ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase zeta; DAGK5 DAGK6DAG kinase zeta DGK-ZETA diacylglycerol kinase zeta diacylglycerol kinase zeta diacylglycerol kinase zeta 104kDa Diglyceride kinase zeta EC 2.7.1.107 hDGKzeta; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase zeta
Gene Aliases: DAGK5; DAGK6; DGK-ZETA; DGKZ; hDGKzeta
UniProt ID: (Human) Q13574
Entrez Gene ID: (Human) 8525
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.