R&D Systems
Manufacturer Code:AF6099SP
Catalog # AF6099SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DGK-zeta Gln800-Val945 Accession # Q13574 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DAG kinase zeta; DAGK5 DAGK6DAG kinase zeta DGKZ DGKzeta DGK-ZETA diacylglycerol kinase zeta diacylglycerol kinase zeta diacylglycerol kinase zeta 104kDa Diglyceride kinase zeta EC 2.7.1.107 hDGKzeta; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase zeta
Gene Aliases: DAGK5; DAGK6; DGK-ZETA; DGKZ; hDGKzeta
UniProt ID: (Human) Q13574
Entrez Gene ID: (Human) 8525
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.