Novus Biologicals
Manufacturer Code:NBP270547
Catalog # NBP270547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A11 |
Immunogen | Full length human recombinant protein of human DGKA(NP_001336) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: 80 kDa diacylglycerol kinase; DAG kinase alpha; DAG kinase alpha DAGK DAGK1diacylglycerol kinase alpha (80kD) DGK-alphaEC 2.7.1.107 diacylglycerol kinase alpha diacylglycerol kinase alpha 80kDa Diglyceride kinase alpha MGC1282180 kDa diacylglycerol kinase MGC42356; DGK-alpha; Diacylglycerol kinase alpha; diacylglycerol kinase, alpha 80kDa; Diglyceride kinase alpha
Gene Aliases: DAGK; DAGK1; DGK-alpha; DGKA
UniProt ID: (Human) P23743
Entrez Gene ID: (Human) 1606
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.