Novus Biologicals
Manufacturer Code:NBP130116
Catalog # NBP130116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antigen maps to a region between residue 400 and 450 of human DiGeorge syndrome critical region gene 8 using the numbering given in entry NP_073557.3 (GeneID 54487). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C22orf12 DGCRK6chromosome 22 open reading frame 12 DiGeorge syndrome critical region 8 DiGeorge syndrome critical region gene 8 Gy1 microprocessor complex subunit DGCR8 pasha; DiGeorge syndrome critical region 8; DiGeorge syndrome critical region gene 8; Microprocessor complex subunit DGCR8
Gene Aliases: C22orf12; DGCR8; DGCRK6; Gy1; LP4941; pasha
UniProt ID: (Human) Q8WYQ5
Entrez Gene ID: (Human) 54487
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.