R&D Systems
Manufacturer Code:MAB10443SP
Catalog # MAB10443SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1019908 |
Immunogen | Chinese Hamster Ovary cell line CHO-derived human DGCR2 Glu22-Ala349 Accession &num P98153 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DGS-C DiGeorge syndrome critical region gene 2 IDDDKFZp686I1730 integral membrane protein deleted in DiGeorge syndrome integral membrane protein DGCR2/IDD KIAA0163 KIAA0163DiGeorge syndrome critical region protein 2 LAN SEZ-12; DiGeorge syndrome critical region protein 2; integral membrane protein deleted in DiGeorge syndrome; Integral membrane protein DGCR2/IDD
Gene Aliases: DGCR2; DGS-C; IDD; KIAA0163; LAN; SEZ-12
UniProt ID: (Human) P98153
Entrez Gene ID: (Human) 9993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.