Novus Biologicals
Manufacturer Code:NBP159220
Catalog # NBP159220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DGCR2(DiGeorge syndrome critical region gene 2) The peptide sequence was selected from the middle region of DGCR2. Peptide sequence AESCYEKSSFLCKRSQTCVDIKDNVVDEGFYFTPKGDDPCLSCTCHGGEP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DGS-C DiGeorge syndrome critical region gene 2 IDDDKFZp686I1730 integral membrane protein deleted in DiGeorge syndrome integral membrane protein DGCR2/IDD KIAA0163DiGeorge syndrome critical region protein 2 LAN SEZ-12; DiGeorge syndrome critical region protein 2; integral membrane protein deleted in DiGeorge syndrome; Integral membrane protein DGCR2/IDD
Gene Aliases: DGCR2; DGS-C; IDD; KIAA0163; LAN; SEZ-12
UniProt ID: (Human) P98153
Entrez Gene ID: (Human) 9993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.