Abnova
Manufacturer Code:PAB17692
Catalog # 89-119-355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human DGAT2L6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DGAT2L6.
For Research Use Only
Protein Aliases: DC3; diacylglycerol O-acyltransferase 2-like 6; Diacylglycerol O-acyltransferase 2-like protein 6; Diacylglycerol O-acyltransferase candidate 3; FLJ25989; hDC3
Gene Aliases: DC3; DGAT2L6
UniProt ID: (Human) Q6ZPD8
Entrez Gene ID: (Human) 347516
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.