Invitrogen
Manufacturer Code:PA575616
Catalog # PIPA575616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human DGAT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA retinol O-fatty-acyltransferase; ARAT; diacylglycerol acyltransferase 2; Diacylglycerol O-acyltransferase 2; Diacylglycerol O-acyltransferase 2 Acyl-CoA retinol O-fatty-acyltransferase ARAT Retinol O-fatty-acyltransferase Diglyceride acyltransferase 2 DGAT2 HMFN1045 UNQ738/PRO1433; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; Diglyceride acyltransferase 2; retinol O-fatty-acyltransferase
Gene Aliases: 0610010B06Rik; ARAT; DGAT-2; DGAT2; GS1999FULL; HMFN1045; UNQ738/PRO1433
UniProt ID: (Human) Q96PD7, (Rat) Q8K4Y4, (Mouse) Q9DCV3
Entrez Gene ID: (Human) 84649, (Rat) 252900, (Mouse) 67800
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.