Invitrogen
Manufacturer Code:PA569667
Catalog # PIPA569667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human DGAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA retinol O-fatty-acyltransferase; ARAT; DGAT2 diacylglycerol O-acyltransferase homolog 2 (mouse) DKFZp686A15125 HMFN1045 GS1999full diacylglycerol O-acyltransferase homolog 2 diacylglycerol O-acyltransferase-like protein 2 isoform 1; diacylglycerol acyltransferase 2; Diacylglycerol O-acyltransferase 2; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; Diglyceride acyltransferase 2; putative diacylglycerol O-acyltransferase; retinol O-fatty-acyltransferase
Gene Aliases: 0610010B06Rik; ARAT; DGAT-2; DGAT2; GS1999FULL; HMFN1045; UNQ738/PRO1433
UniProt ID: (Bovine) Q70VZ8, (Human) Q96PD7, (Rat) Q8K4Y4, (Mouse) Q9DCV3
Entrez Gene ID: (Bovine) 404129, (Dog) 485185, (Rabbit) 100354838, (Horse) 100064387, (Goat) 102174409, (Human) 84649, (Rat) 252900, (Mouse) 67800, (Guinea pig) 100718670
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.