Novus Biologicals
Manufacturer Code:NBP171701AF647
Catalog # NBP171701Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7] |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA retinol O-fatty-acyltransferase; ARAT; Diacylglycerol O-acyltransferase 2; diacylglycerol O-acyltransferase 2 diacylglycerol O-acyltransferase homolog 2 diacylglycerol O-acyltransferase homolog 2 (mouse) diacylglycerol O-acyltransferase-like protein 2 Diglyceride acyltransferase 2 DKFZp686A15125 EC 2.3.1 EC 2.3.1.20 GS1999FULL; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; Diglyceride acyltransferase 2
Gene Aliases: ARAT; DGAT2; GS1999FULL; HMFN1045; UNQ738/PRO1433
UniProt ID: (Human) Q96PD7
Entrez Gene ID: (Human) 84649
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.