R&D Systems
Manufacturer Code:AF7256
Catalog # AF7256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DGAT2 Arg268-Phe377 Accession # Q96PD7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA retinol O-fatty-acyltransferase; ARAT; Diacylglycerol O-acyltransferase 2; diacylglycerol O-acyltransferase 2 diacylglycerol O-acyltransferase homolog 2 diacylglycerol O-acyltransferase homolog 2 (mouse) diacylglycerol O-acyltransferase-like protein 2 Diglyceride acyltransferase 2 DKFZp686A15125 EC 2.3.1 EC 2.3.1.20 GS1999FULL; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; Diglyceride acyltransferase 2
Gene Aliases: ARAT; DGAT2; GS1999FULL; HMFN1045; UNQ738/PRO1433
UniProt ID: (Human) Q96PD7
Entrez Gene ID: (Human) 84649
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.