Invitrogen
Manufacturer Code:PA518634
Catalog # PIPA518634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QNSMKPFKDMDYS) corresponding to the internal amino acids of DGAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT related gene product 1; ACAT-related gene product 1; acyl coenzyme A:cholesterol acyltransferase related gene 1; Acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; ARAT; DGAT1 diacylglycerol O-acyltransferase homolog 1 (mouse) ARGP1 DGAT ACAT related gene product 1 acyl coenzyme A:cholesterol acyltransferase related gene 1 diacylglycerol O-acyltransferase 1 diglyceride acyltransferase; diacylglycerol acyltransferase; Diacylglycerol O-acyltransferase 1; Diglyceride acyltransferase; retinol O-fatty-acyltransferase
Gene Aliases: AGRP1; ARAT; ARGP1; DGAT; DGAT1; DIAR7
UniProt ID: (Human) O75907, (Rat) Q9ERM3
Entrez Gene ID: (Human) 8694, (Rat) 84497
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.