Invitrogen
Manufacturer Code:PA5115115
Catalog # PIPA5115115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DGAT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT related gene product 1; ACAT related gene product 1 ACAT-related gene product 1 Acyl coenzyme A acyl coenzyme A:cholesterol acyltransferase related gene 1 acyl-CoA retinol O-fatty-acyltransferase acyl-CoA:diacylglycerol acyltransferase acyl-coenzyme A:diacylglycerol acyltransferase 1 AGRP1 ARAT ARGP1 C75990 D15Ertd23e Dgat Dgat1 diacylglycerol acyltransferase diacylglycerol acyltransferase 1 diacylglycerol O-acyltransferase 1 DIAR7 Diglyceride acyltransferase EC 2.3.1.20 retinol O-fatty-acyltransferase; ACAT-related gene product 1; acyl coenzyme A:cholesterol acyltransferase related gene 1; Acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; ARAT; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; Diacylglycerol O-acyltransferase 1; Diglyceride acyltransferase; retinol O-fatty-acyltransferase
Gene Aliases: AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; DGAT; DGAT1; DIAR7
UniProt ID: (Human) O75907, (Rat) Q9ERM3, (Mouse) Q9Z2A7
Entrez Gene ID: (Human) 8694, (Rat) 84497, (Mouse) 13350
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.