Invitrogen
Manufacturer Code:PA116985
Catalog # 01-670-138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 200-300 of human DGAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A Diacylglycerol O-acyltransferase 1 EC 2.3.1.20 Diglyceride acyltransferase ACT-related gene product 1; acyl coenzyme A:cholesterol acyltransferase related gene 1; Acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; ARAT; DGAT1B; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; Diacylglycerol O-acyltransferase 1; diacylglycerol O-acyltransferase homolog 1; diacylglycerol O-acyltransferase homolog 1a; Diglyceride acyltransferase; putative diacylglycerol acyltransferase 1; retinol O-fatty-acyltransferase
Gene Aliases: AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; DGAT; DGAT1; DIAR7; wu:fd36g11; zgc:77691
UniProt ID: (Human) O75907, (Bovine) Q8MK44, (Rat) Q9ERM3, (Mouse) Q9Z2A7
Entrez Gene ID: (Human) 8694, (Bovine) 282609, (Rat) 84497, (Mouse) 13350, (Zebrafish) 325875
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.