Abnova
Manufacturer Code:H00008694K
Catalog # 89-209-704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human DGAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human DGAT1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ACAT related gene product 1; ACAT-related gene product 1; acyl coenzyme A:cholesterol acyltransferase related gene 1; Acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; ARAT; ARGP1 DGAT; Diacylglycerol O-acyltransferase 1; Diglyceride acyltransferase
Gene Aliases: AGRP1; ARAT; ARGP1; DGAT; DGAT1; DIAR7
UniProt ID: (Human) O75907
Entrez Gene ID: (Human) 8694
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.