Proteintech
Manufacturer Code:130751AP150UL
Catalog # 50-172-8469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DFNA5 Fusion Protein Ag3746 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deafness autosomal domit 5 DFNA5 GSDME ICERE 1 ICERE1; deafness, autosomal dominant 5; deafness, autosomal dominant 5 homolog; Gasdermin-E; Gasdermin-E, C-terminal; Gasdermin-E, N-terminal; GSDME-CT; GSDME-NT; ICERE-1; Inversely correlated with estrogen receptor expression 1; Non-syndromic hearing impairment protein 5; Non-syndromic hearing impairment protein 5 homolog; nonsyndromic hearing impairment protein
Gene Aliases: 2310037D07Rik; 4932441K13Rik; DFNA5; Dfna5h; EG14210; Fin15; GSDME; ICERE-1; ICERE1
UniProt ID: (Human) Q9UBV3, (Mouse) Q9Z2D3
Entrez Gene ID: (Human) 1687, (Mouse) 54722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.