Invitrogen
Manufacturer Code:PA519891
Catalog # PIPA519891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to 17 amino acids near the center of murine CAD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAD; Caspase-activated deoxyribonuclease; caspase-activated DNase; DFF-40; DNA fragmentation factor 40 kD beta polypeptide (caspase-activated DNase); DNA fragmentation factor 40 kDa subunit; DNA fragmentation factor subunit beta; DNA fragmentation factor, 40 kD, beta polypeptide (caspase-activated DNase); DNA fragmentation factor, 40 kD, beta subunit; DNA fragmentation factor, beta polypeptide (caspase-activated DNase); DNA fragmentation factor, beta subunit; DNase inhibited by DNA fragmentation factor; RP23-254N4.2 40kDa 5730477D02Rik CAD CPAN DFF40 Didff DFF-40 DNA fragmentation factor 40 kDa subunit DNA fragmentation factor subunit beta DNA fragmentation factor 40 kD beta subunit DNase inhibited by DNA fragmentation factor caspase-activated DNase caspase-activated deoxyribonuclease
Gene Aliases: 40kDa; 5730477D02Rik; CAD; CPAN; DFF40; Dffb; Didff
UniProt ID: (Rat) Q99N34, (Mouse) O54788
Entrez Gene ID: (Rat) 84359, (Mouse) 13368
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.