Novus Biologicals
Manufacturer Code:NBP177010SS
Catalog # NBP177010SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to amino acids near the amino terminus of mouse ICAD. The immunogen is located within the first 50 amino acids of ICAD. Amino Acid Squence: ELSRGASAPDPDDVRPLKPC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: DFF-45; DFF1DNA fragmentation factor 45 kDa subunit DFF45DNA fragmentation factor subunit alpha DFF-45DNA fragmentation factor 45 kD alpha polypeptide DNA fragmentation factor 45kDa alpha polypeptide ICAD Inhibitor of CAD; DFF45; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; DNA fragmentation factor, 45kDa, alpha polypeptide; ICAD; Inhibitor of CAD
Gene Aliases: DFF-45; DFF1; DFF45; DFFA; H13; ICAD
UniProt ID: (Human) O00273
Entrez Gene ID: (Human) 1676
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.