R&D Systems
Manufacturer Code:AF4679
Catalog # AF4679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DFF45 Met1-Thr331 Accession # O00273 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DFF-45; DFF1 DFF1DNA fragmentation factor 45 kDa subunit DFF45DNA fragmentation factor subunit alpha DFF-45DNA fragmentation factor 45 kD alpha polypeptide DFFA DNA fragmentation factor 45kDa alpha polypeptide ICAD Inhibitor of CAD; DFF45; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; DNA fragmentation factor, 45kDa, alpha polypeptide; ICAD; Inhibitor of CAD
Gene Aliases: DFF-45; DFF1; DFF45; DFFA; H13; ICAD
UniProt ID: (Human) O00273
Entrez Gene ID: (Human) 1676
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.