Novus Biologicals
Manufacturer Code:NBP2828380.1ML
Catalog # NB030767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human DERA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-deoxy-D-ribose 5-phosphate aldolase; 2-deoxyribose-5-phosphate aldolase homolog; Deoxyriboaldolase; deoxyriboaldolase deoxyribose-phosphate aldolase (putative) phosphodeoxyriboaldolase; Deoxyribose-phosphate aldolase; deoxyribose-phosphate aldolase (putative); DERA; Phosphodeoxyriboaldolase; putative deoxyribose-phosphate aldolase
Gene Aliases: CGI-26; DEOC; DERA
UniProt ID: (Human) Q9Y315
Entrez Gene ID: (Human) 51071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.