Invitrogen
Manufacturer Code:PA525343
Catalog # PIPA525343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 10-38 amino acids from the N-terminal region of human DEPDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle control protein SDP35; DEP domain-containing protein 1A; DEP.8 DEPDC1-V2 DEPDC1A SDP35 5830484J08Rik DEP domain-containing protein 1A cell cycle control protein SDP35
Gene Aliases: DEP.8; DEPDC1; DEPDC1-V2; DEPDC1A; SDP35
UniProt ID: (Human) Q5TB30
Entrez Gene ID: (Human) 55635
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.