Abnova
Manufacturer Code:H00055635A01
Catalog # 89-107-052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 93 a.a. to 186 a.a) partial recombinant protein with GST tag.,DEPDC1 (NP_060249 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant DEPDC1.
For Research Use Only
Protein Aliases: cell cycle control protein SDP35; DEP domain-containing protein 1A; DEP.8; DEPDC1-V2; DEPDC1A; FLJ20354; SDP35
Gene Aliases: DEP.8; DEPDC1; DEPDC1-V2; DEPDC1A; SDP35
UniProt ID: (Human) Q5TB30
Entrez Gene ID: (Human) 55635
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.