Abnova
Manufacturer Code:H00221120A01
Catalog # 89-107-709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 139 a.a) full-length recombinant protein with GST tag.,DEPC-1 (AAH15155 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant DEPC-1.
For Research Use Only
Protein Aliases: ABH3; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; Alkylated DNA repair protein alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC-1; DEPC1; hABH3; MGC118790; MGC118792; MGC118793; PCA1; Prostate cancer antigen 1; prostate cancer antigen-1
Gene Aliases: ABH3; ALKBH3; DEPC-1; DEPC1; hABH3; PCA1
UniProt ID: (Human) Q96Q83
Entrez Gene ID: (Human) 221120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.