Invitrogen
Manufacturer Code:PA553255
Catalog # PIPA553255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DENND4C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DENN domain-containing protein 4C; DENN/MADD domain containing 4A pseudogene; DENN/MADD domain containing 4C; likely ortholog of H. sapiens chromosome 9 open reading frame 55 (C9orf55)
Gene Aliases: 1700065A05Rik; AA420392; bA513M16.3; C9orf55; C9orf55B; DENND4C; RAB10GEF; RGD1308489
UniProt ID: (Human) Q5VZ89, (Mouse) A6H8H2
Entrez Gene ID: (Human) 55667, (Mouse) 329877, (Rat) 313340
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.