Novus Biologicals
Manufacturer Code:NBP179530
Catalog # NBP179530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DENND2CThe immunogen for this antibody is DENND2C. Peptide sequence DIFESKRGKKKVKLHSYTGKELPPTKGETSGNESDAEYLPKNRHKRLAQL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DENN domain-containing protein 2C; DENN domain-containing protein 2C DENN/MADD domain containing 2C DKFZp686N1631; DENN/MADD domain containing 2C; RP5-1156J9.1
Gene Aliases: DENND2C; dJ1156J9.1
UniProt ID: (Human) Q68D51
Entrez Gene ID: (Human) 163259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.