Novus Biologicals
Manufacturer Code:NBP169231
Catalog # NBP169231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DENND1B(DENN/MADD domain containing 1B) The peptide sequence was selected from the N terminal of DENND1B. Peptide sequence YKLLNTLADYLAKELENDLNETLRSLYNHPVPKANTPVNLSVNQEIFIAC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1ORF18 C1orf218 chromosome 1 open reading frame 218 Connecdenn 2 DENN domain-containing protein 1B DENN/MADD domain containing 1B DKFZp547O0715 FAM31BMGC103810 FLJ20054 FLJ21179 FLJ33888 member B Protein FAM31B; Connecdenn 2; DENN domain-containing protein 1B; DENN/MADD domain containing 1B; family with sequence similarity 31, member B; Protein FAM31B
Gene Aliases: C1ORF18; C1orf218; DENND1B; FAM31B
UniProt ID: (Human) Q6P3S1
Entrez Gene ID: (Human) 163486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.