Invitrogen
Manufacturer Code:PA570324
Catalog # PIPA570324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human DENND1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Connecdenn; Connecdenn 1; DENN domain-containing protein 1A; DENN domain-containing protein 1A pseudogene; DENN/MADD domain containing 1A; FAM31A KIAA1608 RP11-230L22.3 DENN domain-containing protein 1A connecdenn 1; Protein FAM31A
Gene Aliases: 6030446I19Rik; connecdenn; DENND1A; FAM31A; KIAA1608; RGD1307927
UniProt ID: (Human) Q8TEH3, (Mouse) Q8K382
Entrez Gene ID: (Horse) 100067362, (Rabbit) 100340700, (Human) 57706, (Dog) 609706, (Pig) 102166001, (Bovine) 514134, (Guinea pig) 100735930, (Rat) 311913, (Mouse) 227801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.