Abnova
Manufacturer Code:PAB21307
Catalog # 89-122-798
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human DENND1A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant DENND1A.
For Research Use Only
Protein Aliases: Connecdenn; Connecdenn 1; DENN domain-containing protein 1A; DENN/MADD domain containing 1A; FAM31A FLJ38464 KIAA1608 RP11-230L22.3; Protein FAM31A
Gene Aliases: DENND1A; FAM31A; KIAA1608
UniProt ID: (Human) Q8TEH3
Entrez Gene ID: (Human) 57706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.