Novus Biologicals
Manufacturer Code:NBP2828330.1ML
Catalog # NB030762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human DEM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf176 chromosome 1 open reading frame 176 defects in morphology 1 homolog (S. cerevisiae) Defects in morphology protein 1 homolog EC 3.1.11.- Exo V FLJ11445 FLJ13183 FLJ21144; defects in morphology 1 homolog; Defects in morphology protein 1 homolog; Exo V; Exonuclease V; probable exonuclease V
Gene Aliases: C1orf176; DEM1; Exo V; EXO5; hExo5
UniProt ID: (Human) Q8WTW9
Entrez Gene ID: (Human) 64789
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.